Learn More
Hach Company Thermometer with Probe
Supplier:  Hach Company 2630605
Encompass
Small light weight and inexpensive these units provide extremely accurate and reliable temperature measurement with all the versatility and range of type K thermometers Features relative min/max/ave temperature hold and touch tone buttons Results are displayed on a large 1 in x 1 in LCD An adjustment allows the user to easily calibrate to ice point or any other standard Comes ready to use with a 9V battery and beaded wire probe (Please Note Unit with certification is PN 2630600)
Catalog No. 50-002-57126