Learn More
Thermco Products Inc NIST Traceable - Factory Certified Non-Mercury Glass Thermometers - Spirit Filled
Catalog No. NC1577795
Encompass_Preferred
Range: -1/201°C | Div: 0.2°C | Length: 610mm
Catalog No. NC1577795 Supplier Thermco Products Inc Supplier No. ACC1007SFC
Range: -1/201°C | Div: 0.2°C | Length: 610mm