Learn More
Thermco Products Inc General lab Non-Hg thermometer, Spirit, -20 deg./150 deg.C & 0 deg./300 deg.F, 76mm imm
Catalog No. NC0605679
Encompass_Preferred
Spirit filled double scale thermometer, white back tubing, with toughened bulbs. Suspension ring for hanging, vertical numbering. Individual serial numbered. Packed in non-roll square tubing. Permanent markings.
Catalog No. NC0605679 Supplier Thermco Products Inc Supplier No. B150C300FW3S
Spirit filled double scale thermometer, white back tubing, with toughened bulbs. Suspension ring for hanging, vertical numbering. Individual serial numbered. Packed in non-roll square tubing. Permanent markings.