Learn More
Ptc Instruments THERMOMETER, SURFACE, 50-750F
Catalog No. NC0909040
Encompass
Designed for applications such as: hot plates; griddles; die-cast dies; foundry core boxes; annealing ovens; weld preheats; motors, generators, and engines; bearings; boilers, tanks ect. •Accuracy of ±2% of Full Scale Range •NIST/A2LA ISO 17025 3 Point Certification Available •Every Unit Individually Calibrated •Time constant from 0.06 to 1 minute (depending on the range) •Diameter 2 in. (5.1 cm). •Height ½ " (1.3 cm)
Catalog No. NC0909040 Supplier Ptc Instruments Supplier No. 314F
Designed for applications such as: hot plates; griddles; die-cast dies; foundry core boxes; annealing ovens; weld preheats; motors, generators, and engines; bearings; boilers, tanks ect. •Accuracy of ±2% of Full Scale Range •NIST/A2LA ISO 17025 3 Point Certification Available •Every Unit Individually Calibrated •Time constant from 0.06 to 1 minute (depending on the range) •Diameter 2 in. (5.1 cm). •Height ½ " (1.3 cm)