Learn More
Thermco Products Inc Special application Hg, 0 deg./300 deg.C, 1.0C div, 125mm imm, 405mm
Catalog No. NC0630050
Encompass_Preferred
Precision mercury thermometer designed to be used in laboratory ovens. The immersion is 125mm. There is a stem enlargement at the 125mm immersion mark, so the thermometer is immersed correctly. The thermometer is also used in high temperature baths. It is accurate to N.I.S.T. tolerance and can be certified and taceable to N.I.S.T. Individually serialized
Catalog No. NC0630050 Supplier Thermco Products Inc Supplier No. ACC629
Precision mercury thermometer designed to be used in laboratory ovens. The immersion is 125mm. There is a stem enlargement at the 125mm immersion mark, so the thermometer is immersed correctly. The thermometer is also used in high temperature baths. It is accurate to N.I.S.T. tolerance and can be certified and taceable to N.I.S.T. Individually serialized