Learn More
Cole-Parmer Reversible 48-Point Vertical Magnetic Temperature Label, 26-120F; 10/Pk
Catalog No. 501961472 Shop All Cole-Parmer Products
Encompass_Preferred
These self-adhesive reusable temperature labels provide a safe, accurate, easy-to-read means for monitoring temperatures. Indicating strips use liquid crystal technology to display the current temperature at an economical cost. The strip will turns a green color when ambient temperature is reached. Shelf life: 1 year after date of purchase.
Catalog No. 50-196-1472 Supplier Cole-Parmer Supplier No. 0807848
These self-adhesive reusable temperature labels provide a safe, accurate, easy-to-read means for monitoring temperatures. Indicating strips use liquid crystal technology to display the current temperature at an economical cost. The strip will turns a green color when ambient temperature is reached. Shelf life: 1 year after date of purchase.