Learn More
Med Vet International McKesson Refrigerator / Freezer Thermometer, Digital Display, -58° to +158°F (-50° to +70°C), MCK895RFV, Each
Catalog No. 503046370
Encompass_Preferred
Digital Refrigerator / Freezer Thermometer with Alarm McKesson Fahrenheit / Celsius -58° to +158°F (-50° to +70°C) Glycol Bottle Probe / Internal Sensor Multiple Mounting Options Battery Operated Triple display shows current, min/max temperatures with a 10-second refresh rate, accuracy of ±0.5° C and a 0.1° display resolution Certified, serialized and traceable and has been tested against a standard calibrated at the National Institute of Standards and Technology (NIST) Monitors refrigerators or freezer temperatures from -40 to 158°F (-40 - 70° C ) Audible alarm indicates when the programmable low and high limits set are breached Optional display formats for time, date, °C / °F Fold-away stand, magnet and hook-and-loop attachments allow multiple mounting options
Catalog No. 50-304-6370 Supplier Med Vet International Supplier No. 1074440EA
Digital Refrigerator / Freezer Thermometer with Alarm McKesson Fahrenheit / Celsius -58° to +158°F (-50° to +70°C) Glycol Bottle Probe / Internal Sensor Multiple Mounting Options Battery Operated Triple display shows current, min/max temperatures with a 10-second refresh rate, accuracy of ±0.5° C and a 0.1° display resolution Certified, serialized and traceable and has been tested against a standard calibrated at the National Institute of Standards and Technology (NIST) Monitors refrigerators or freezer temperatures from -40 to 158°F (-40 - 70° C ) Audible alarm indicates when the programmable low and high limits set are breached Optional display formats for time, date, °C / °F Fold-away stand, magnet and hook-and-loop attachments allow multiple mounting options