Learn More
MSC Fibre-Metal Cap Style Hard Hat, Fiberglass, ANSI Type I, Class G, 1 ea
Catalog No. 5000507944
Encompass
Engineered for strength and durability, this cap-style hard hat is constructed from a proprietary injection-molded fiberglass compound that is eight times stronger than standard high-density polyethylene. The shell provides exceptional impact and crack resistance and maintains its structural integrity in extreme heat up to 500°F. It features a permanent, no-chip, no-peel solid color resin finish and is factory-assembled with a non-slip, non-strip ratchet suspension system for a secure and comfortable fit.
  • Injection-molded fiberglass compound for superior strength
  • Heat resistance up to 500°F
  • Smooth, matte finish
  • Includes non-slip, non-strip ratchet headgear
  • Compatible with Quick-Lok® and Speedy® Loop attachment systems
  • SEI certified to meet ANSI Z89.1-2009 Type I Class G and CSA Z94.1-2005 Type I Class G or C standards
Catalog No. 50-005-07944 Supplier MSC Supplier No. 04285045
Engineered for strength and durability, this cap-style hard hat is constructed from a proprietary injection-molded fiberglass compound that is eight times stronger than standard high-density polyethylene. The shell provides exceptional impact and crack resistance and maintains its structural integrity in extreme heat up to 500°F. It features a permanent, no-chip, no-peel solid color resin finish and is factory-assembled with a non-slip, non-strip ratchet suspension system for a secure and comfortable fit.
  • Injection-molded fiberglass compound for superior strength
  • Heat resistance up to 500°F
  • Smooth, matte finish
  • Includes non-slip, non-strip ratchet headgear
  • Compatible with Quick-Lok® and Speedy® Loop attachment systems
  • SEI certified to meet ANSI Z89.1-2009 Type I Class G and CSA Z94.1-2005 Type I Class G or C standards