Learn More
New England Biolabs, Inc. ThermoPol Reaction Buffer Pack – 6 ml (4 x 1.5 ml)
SDP
Catalog No. 50811565
Encompass
10X ThermoPol Reaction Buffer is optimized for use with Vent and Deep Vent DNA Polymerases. This buffer also provides superior reaction conditions for other thermophilic DNA polymerases, including Taq DNA Polymerase, as well as various other DNA and RNA modifying enzymes. Supplemental MgSO4 is provided to maximize flexibility for users that require additional Mg2+ for their specific application. See individual product descriptions for the recommended buffer for each enzyme, or visit the FAQ page for this product on the NEB web site for a complete list of enzymes for which this buffer is either recommended or compatible.
Catalog No. 50-811-565 Supplier New England Biolabs, Inc. Supplier No. B9004S
10X ThermoPol Reaction Buffer is optimized for use with Vent and Deep Vent DNA Polymerases. This buffer also provides superior reaction conditions for other thermophilic DNA polymerases, including Taq DNA Polymerase, as well as various other DNA and RNA modifying enzymes. Supplemental MgSO4 is provided to maximize flexibility for users that require additional Mg2+ for their specific application. See individual product descriptions for the recommended buffer for each enzyme, or visit the FAQ page for this product on the NEB web site for a complete list of enzymes for which this buffer is either recommended or compatible.