Learn More
Metrohm USA THERMOPROBE FOR 859 TITROTHERM
SDP
Catalog No. NC0434356 Shop All Metrohm USA Products
Encompass
Temperature sensor for thermometric titration with the 859 Titrotherm. For aqueous or nonaqueous solutions.
Catalog No. NC0434356 Supplier Metrohm USA Supplier No. 69011020
Temperature sensor for thermometric titration with the 859 Titrotherm. For aqueous or nonaqueous solutions.