Learn More
AdipoGen Thermorubin
SDP
Supplier:  AdipoGen AGCN20339M005
Encompass_Preferred
Chemical. CAS 37577-75-6. Formula C32H24O12. MW 600.5. Isolated from Thermoactinomyces sp. Anthracene antibiotic. Antibacterial agent. Active against both Gram-positive and Gram-negative bacteria. Inhibits the initiation stage of bacterial protein synthesis by binding to the inter-subunit bridge B2a on the 70S ribosome. Does not inhibit DNA and RNA synthesis. Potent aldose reductase inhibitor.
Catalog No. 50-256-2564