Learn More
Sonoco™ ThermoSafe Absorbant Pad for Model 470 Diagnostic Shipper
Catalog No. 03530203 Shop All Sonoco ThermoSafe Products
Change view
Click to view available options
Quantity:
50/Cs.
Catalog No. Quantity
03-530-203 50/Cs.
1 options
Catalog No. 03-530-203 Supplier Sonoco™ Supplier No. 471PAD50
Add to Cart
Add to Cart
Offers absorbent material for maximum capacity of liquid
  • Meets requirements for the transport of diagnostic specimens
  • For use with tubes or bags
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50/Cs.
For Use With (Equipment) For ThermoSafe diagnostic shipper Model 470
Applications Diagnostic, Infectious, Lab and Medical
Type Absorbent Pad
Duration Transport Short-Term