Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sonoco™ ThermoSafe Boston Round Bottle Mailer

Catalog No. 0352531 Shop All Sonoco ThermoSafe Products
Change view
Click to view available options
Type:
Mailer Model 396
Quantity:
100/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
03-525-31 Mailer Model 396 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-525-31 Supplier Sonoco™ Supplier No. 395
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lightweight, economical EPS shippers provide maximum breakage protection for known non-hazardous materials.

  • Lightweight polystyrene foam construction
  • Reusable and recyclable

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100/Pk.
For Use With (Equipment) VacuTainer, Microtainer, and other collection/transport tubes
Applications Maximum breakage protection for shipping known non-hazardous materials
Type Mailer Model 396
Dimensions (L x W x H) Exterior 4.37 x 3.25 x 3.25 in.
Diameter (English) Exterior 3.25 in.
Diameter (English) Interior 2 in.
Description Foam only lab mailer
Content And Storage Boston Round or aluminum can
Capacity 57 mL (2 oz.)
Material Polystyrene Foam
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.