Learn More
Sonoco™ ThermoSafe Diagnostic Shipper, Eight Tube Mailing System: Foam Mailer
Catalog No. 03527B Shop All Sonoco ThermoSafe Products
Change view
Click to view available options
Includes:
Safety Mailer (EPS Foam Unit Only)
Catalog No. Includes
03-527B Safety Mailer (EPS Foam Unit Only)
1 options
Catalog No. 03-527B Supplier Sonoco™ Supplier No. 470
Add to Cart
Add to Cart
Includes
Cat. No. 03-527B includes eight-tube safety mailer only. Complete set (Cat. No. 03-527C) includes: 10mL vial tubes, foam mailer with lid, absorbent material, water-resistant tape, pressure-lock plastic bag, 200 lb. test corrugated shipping carton with antitamper locking tabs and instructions.
Leak-resistant, EPS foam mailer holds 30mL bottle, eight 10mL tubes, materials for insurance testing and other paperwork. (Note: this includes the foam unit only. For the complete set including tubes, tape, and shipping carton, see 03-527C.)
Leak-resistant, EPS foam mailer holds 30mL bottle, eight 10mL tubes, materials for insurance testing and other paperwork.
  • Tight-fitting plug cover accepts required absorbent material
  • Validated for the transport of diagnostic samples
Compliance
Meets U.S. Postal Regulations, DOT regulations, and IATA Packaging Instruction 650; tubes and shipping system validated for the transport of diagnostic samples
TRUSTED_SUSTAINABILITY
Specifications
Includes Safety Mailer (EPS Foam Unit Only)
Certifications/Compliance Meets U.S. Postal Regulations, DOT regulations, and IATA Packaging Instruction 650; tubes and shipping system validated for the transport of diagnostic samples
Applications Diagnostic materials
Material Expanded Polystyrene Foam
WARNING: Reproductive Harm - www.P65Warnings.ca.gov