Learn More
Sonoco™ ThermoSafe LabMailer Shipping Containers
Catalog No. p-150521 Shop All Sonoco ThermoSafe Products
Change view
Click to view available options
Type:
Mailer Model 312
Mailer Model 402
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
03-525-34B Mailer Model 402
03-531 Mailer Model 312
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-525-34B Supplier Sonoco™ Supplier No. 402
Only null left
Add to Cart
Add to Cart
Accommodates a variety of tubes, vials, cans and bottles
  • 03-525-23B: Mailing sleeve for Model 03-525-34B
  • 03-525-34B: Sturdy 0.5 in. (1.3 cm) walls protect against shock and breakage
  • Foam mailer and paperboard mailing sleeve sold separately
  • Interior dimensions: 4L × 3.25W × 3.25 in.H (10.2 × 8.3 × 8.3cm)
TRUSTED_SUSTAINABILITY
Specifications
Applications Maximum breakage protection for shipment of known non-hazardous materials
Type Mailer Model 402
Dimensions (L x W x H) Exterior 5.25 x 4.37 x 4.37 in.
Dimensions (D x W x H) Interior 4 x 3.5 x 3.25 in.
Description Holds Tubes, Vials, cans, bottles. Can be used with poly-pad cushion to keep containers from shifting.
Material Reusable/recyclable polystyrene foam
WARNING: Reproductive Harm - www.P65Warnings.ca.gov