Learn More
Sonoco™ ThermoSafe Medical Transporter Qualified Medical Shippers
Ideal for operating room and blood bank storage and transport of packed red blood cells, tissues, organs, medical devices, pharmaceutical samples, and other temperature-sensitive samples
Supplier: Sonoco™ 960
Includes: Model 930 includes medical transporter with wheels and retractable handle, wire basket, five polyethylene liners, three PolarPack™ rigid bottles (32 oz.), two PolarPack gel packs (16 oz.), and qualification report. Model 960 includes medical transporter, two wire baskets, five polyethylene liners, and qualifications report. Collapsible dolly for model 960 available separately.
Description
- Qualified by ISC Labs
- Nylon exterior
- Polyethylene liner
- Maintain up to eight (model 930) or 16 (model 960) packed red blood cell units at 1° to 6°C while in storage for 6 hours or more
- Hold up to eight (model 930) or 16 (model 960) packed red blood cell units at 1° to 10°C while in transit for up to 24 hours
Specifications
960 medical transporter, 2 wire baskets, 5 polyethylene liners, qualification report | |
Storage and transport of packed red blood cells, tissues, organs, medical devices, pharmaceutical samples, and other temperature-sensitive samples | |
Medical Transporter |