Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sonoco™ ThermoSafe Metalized Bubble Pouches

Catalog No. 03600158 Shop All Sonoco ThermoSafe Products
Change view
Click to view available options
Length (English) Exterior:
8 in.
12 in.
Width (English) Exterior:
11 in.
17 in.
23 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Exterior Width (English) Exterior
03-600-158 12 in. 23 in.
03-600-154 8 in. 11 in.
03-600-156 12 in. 17 in.
03-600-155 12 in. 11 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 03-600-158 Supplier Sonoco™ Supplier No. MP1223
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lightweight, economical EPS shippers provide maximum breakage protection for known non-hazardous materials.

  • Accommodates VacuTainer™, Microtainer™, and other collection/transport tubes
  • Available in multi-tube, slide, and tube/slide combinations
  • Lightweight polystyrene foam construction saves postage
  • Reusable and recyclable
TRUSTED_SUSTAINABILITY

Specifications

Quantity 150 Pouches/Pk.
Length (English) Exterior 12 in.
Width (English) Exterior 23 in.
Includes 2 in. Self sealing lip and tape
Type Bubble Pouch
Dimensions (L x W) 12 x 23 in.
Material 0.187 in. thick metallized bubble
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.