Learn More
Brookfield Engineer Labs THERMOSEL/SSA SPINDLE NOSC4-21
Catalog No. NC9183534
Encompass
Spindle, For use with the Thermosel System, Selection is determined by the viscosity measurement range that is required
Catalog No. NC9183534 Supplier Brookfield Engineer Labs Supplier No. SC421
Spindle, For use with the Thermosel System, Selection is determined by the viscosity measurement range that is required