Learn More
Brookfield Engineer Labs THERMOSEL/SSA SPINDLE OSC4-27
Catalog No. NC9707901
Encompass
Spindle, Entry Code: 27, SMC: 25, SRC: 0.34, Each spindle has a two digit entry code which is entered via the keypad on the DV-II +, The entry codeallows the DV-II + to calculate Viscosity, Shear Rate and Shear Stress values.
Catalog No. NC9707901 Supplier Brookfield Engineer Labs Supplier No. SC427
Spindle, Entry Code: 27, SMC: 25, SRC: 0.34, Each spindle has a two digit entry code which is entered via the keypad on the DV-II +, The entry codeallows the DV-II + to calculate Viscosity, Shear Rate and Shear Stress values.