Learn More
GRANT INSTRUMENTS Thermoshaker for microplates, holds 4 standard microplates, ambient +5°C to +60°C
SDP
Catalog No. NC1507725
Encompass_Preferred
Excellent temperatures uniformity across the platform/microplate (patented bi-directional heating of microplates) combined with variable speed and variable temperature produces the ideal thermoshaker for microplate incubations.Can be used with all types of standard depth microplates and offers three devices in one for maximum versatility and efficiency.A microplate thermoshaker.A compact benchtop incubator without shaking.A microplate shaker in cold or ambient temperatures.Features:Temperature setting range: + 25°C to 100°C.Temperature stability ± 0.1°C, temperature uniformity ± 0.2°C due to the bi-directional heating system (platform and lid).Shaking speed: 250 to 1200rpm.
Catalog No. NC1507725 Supplier GRANT INSTRUMENTS Supplier No. PHMP4
Excellent temperatures uniformity across the platform/microplate (patented bi-directional heating of microplates) combined with variable speed and variable temperature produces the ideal thermoshaker for microplate incubations.Can be used with all types of standard depth microplates and offers three devices in one for maximum versatility and efficiency.A microplate thermoshaker.A compact benchtop incubator without shaking.A microplate shaker in cold or ambient temperatures.Features:Temperature setting range: + 25°C to 100°C.Temperature stability ± 0.1°C, temperature uniformity ± 0.2°C due to the bi-directional heating system (platform and lid).Shaking speed: 250 to 1200rpm.