Learn More
New England Biolabs, Inc. Thermostable FEN1 – 1600 units
SDP
Catalog No. 501526397
Encompass
Thermostable Flap Endonuclease 1, FEN1, catalyzes the cleavage of 5 DNA flaps from branched double stranded DNA substrates, creating a 5' phosphate terminus. FEN1 products can be ligated by DNA ligase to create double stranded DNA. In vivo, FEN1 is an essential component of the Okazaki fragment maturation pathway, and also plays a role in base excision repair.
Catalog No. 50-152-6397 Supplier New England Biolabs, Inc. Supplier No. M0645S
Thermostable Flap Endonuclease 1, FEN1, catalyzes the cleavage of 5 DNA flaps from branched double stranded DNA substrates, creating a 5' phosphate terminus. FEN1 products can be ligated by DNA ligase to create double stranded DNA. In vivo, FEN1 is an essential component of the Okazaki fragment maturation pathway, and also plays a role in base excision repair.