Learn More
New England Biolabs, Inc. Thermostable Inorganic Pyrophosphatase – 250 units
SDP
Catalog No. 50994864
Encompass
Inorganic pyrophosphatase (PPase) catalyzes the hydrolysis of inorganic pyrophosphate to form orthophosphate: P2O7-4 + H2O -> 2HPO4-2.
Catalog No. 50-994-864 Supplier New England Biolabs, Inc. Supplier No. M0296S
Inorganic pyrophosphatase (PPase) catalyzes the hydrolysis of inorganic pyrophosphate to form orthophosphate: P2O7-4 + H2O -> 2HPO4-2.