Learn More
New England Biolabs, Inc. Thermostable OGG - 500 units
SDP
Catalog No. 502336248
Encompass
Thermostable OGG is an archaeal 8-oxoguanine (8oxoG) DNA glycosylase which acts both as aN-glycosylase and an AP-lyase. TheN-glycosylase activity releases the damaged purine (8-oxoguanine) from double stranded DNA, generating an apurinic (AP) site. The AP-lyase activity cleaves 3' to the AP site leaving a 5' phosphate and a 3'-phospho-alpha, beta-unsaturated aldehyde. Unlike some other DNA glycosylases, Thermostable OGG specifically recognizes and cleaves only 8oxoG and no other modified bases.
Catalog No. 50-233-6248 Supplier New England Biolabs, Inc. Supplier No. M0464S
Thermostable OGG is an archaeal 8-oxoguanine (8oxoG) DNA glycosylase which acts both as aN-glycosylase and an AP-lyase. TheN-glycosylase activity releases the damaged purine (8-oxoguanine) from double stranded DNA, generating an apurinic (AP) site. The AP-lyase activity cleaves 3' to the AP site leaving a 5' phosphate and a 3'-phospho-alpha, beta-unsaturated aldehyde. Unlike some other DNA glycosylases, Thermostable OGG specifically recognizes and cleaves only 8oxoG and no other modified bases.