Learn More
New England Biolabs, Inc. Thermostable RNase H – 250 units
SDP
Catalog No. 501527866
Encompass
Thermostable RNase H is an endoribonuclease that is functional at high temperatures and selectively hydrolyzes the phosphodiester bonds of an RNA strand in an RNA:DNA hybrid molecule, while leaving the DNA strand intact. In addition, Thermostable RNase H does not degrade single- or double-stranded RNA or DNA. While the Thermostable and E. coli RNase H homologs both exhibit these same endoribonucleolytic properties, the thermostable enzyme displays activity at higher temperatures with optimal activity above 65C. These properties allow use in applications where higher assay temperatures are desired.
Catalog No. 50-152-7866 Supplier New England Biolabs, Inc. Supplier No. M0523S
Thermostable RNase H is an endoribonuclease that is functional at high temperatures and selectively hydrolyzes the phosphodiester bonds of an RNA strand in an RNA:DNA hybrid molecule, while leaving the DNA strand intact. In addition, Thermostable RNase H does not degrade single- or double-stranded RNA or DNA. While the Thermostable and E. coli RNase H homologs both exhibit these same endoribonucleolytic properties, the thermostable enzyme displays activity at higher temperatures with optimal activity above 65C. These properties allow use in applications where higher assay temperatures are desired.