Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Velp™ Thermostatic Bath
SDP

Catalog No. 11392166
Change view
Click to view available options
Length (Metric):
410 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-392-166 410 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-392-166 Supplier Velp™ Supplier No. A00001055
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Velp™ Thermostatic Bath for use with MULTISTIRRER and MULTISTIRRER DIGITAL.

  • For use with MULTISTIRRER and MULTISTIRRER DIGITAL

Specifications

For Use With (Equipment) MULTISTIRRER and MULTISTIRRER DIGITAL
Height (Metric) 180 mm
Length (Metric) 410 mm
Type Heating Bath for Oil
Warranty 24 Months
Width (Metric) 280 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.