Learn More
Grainger Danfoss Thermostatic Radiator Valve, Angle, Brass, 1 in., 1 ea.
Catalog No. 502763500
Encompass_Preferred
This thermostatic radiator valve features an angle body design with an FNPT inlet and MNPT union tailpiece outlet for use in hot water and low pressure steam heating systems. It is constructed from nickel-plated brass and is designed to remain normally open when no operator is mounted.
  • Replaceable packing gland assembly for maintenance during operation
  • Grease cup for life-long lubrication of the o-ring packing
  • Durable EPDM rubber valve disc for a positive seal
  • Rated for differential pressures up to 15 psi
  • Conforms to ASHRAE / ANSI Standard 102-1983
  • Maximum flow temperature of 120°C (250°F)
  • Flow capacity (Cv) of 2.8
Catalog No. 50-276-3500 Supplier Grainger Supplier No. 161J10
This thermostatic radiator valve features an angle body design with an FNPT inlet and MNPT union tailpiece outlet for use in hot water and low pressure steam heating systems. It is constructed from nickel-plated brass and is designed to remain normally open when no operator is mounted.
  • Replaceable packing gland assembly for maintenance during operation
  • Grease cup for life-long lubrication of the o-ring packing
  • Durable EPDM rubber valve disc for a positive seal
  • Rated for differential pressures up to 15 psi
  • Conforms to ASHRAE / ANSI Standard 102-1983
  • Maximum flow temperature of 120°C (250°F)
  • Flow capacity (Cv) of 2.8