Learn More
Grainger Danfoss RA2000 Radiator Valve, Angle, Brass, 1¼ in.
Catalog No. 502762022
Encompass_Preferred
The RA2000 is a non-electric, thermostatic angle valve designed for radiator applications, featuring a 1¼ in. connection size. Constructed from durable nickel-plated brass, this valve is engineered for use in hot water heating systems, providing reliable flow control with a capacity of 2.8 Cv. Its design includes an integral union tailpiece for efficient installation and maintenance.
  • Sturdy EPDM rubber valve disc provides a positive seal against the valve seat at differential pressures up to 15 psi.
  • Replaceable packing gland assembly allows for maintenance while the system is in operation.
  • Packing gland includes a grease cup to ensure the O-ring packing is lubricated for life.
  • Valve remains normally open when no operator is mounted.
  • Plastic cap protects the valve pushpin and allows for manual control during installation.
Catalog No. 50-276-2022 Supplier Grainger Supplier No. 161J13
The RA2000 is a non-electric, thermostatic angle valve designed for radiator applications, featuring a 1¼ in. connection size. Constructed from durable nickel-plated brass, this valve is engineered for use in hot water heating systems, providing reliable flow control with a capacity of 2.8 Cv. Its design includes an integral union tailpiece for efficient installation and maintenance.
  • Sturdy EPDM rubber valve disc provides a positive seal against the valve seat at differential pressures up to 15 psi.
  • Replaceable packing gland assembly allows for maintenance while the system is in operation.
  • Packing gland includes a grease cup to ensure the O-ring packing is lubricated for life.
  • Valve remains normally open when no operator is mounted.
  • Plastic cap protects the valve pushpin and allows for manual control during installation.