Learn More
Electron Microscopy Sciences Thermotech™ PTFE Beakers 250 ml, 94 mm height, 75 mm O.D.
SDP
Catalog No. 50996290
Encompass_Preferred
The beakers feature a unique combination of a pure "PTFE" Body with a specially formulated stabilized "PTFE" Carbon base as a single molding resulting in a totally inert pure "PTFE" inner surface. It is heatable to 270 degree C without distortion and has improved heat transfer. All "PTFE" Beaker caps can be used for Thermotech "PTFE" Beakers, accordingly to its sizes.
Height: 94 mm
Catalog No. 50-996-290 Supplier Electron Microscopy Sciences Supplier No. 6093025
Add to Cart
Add to Cart
The beakers feature a unique combination of a pure "PTFE" Body with a specially formulated stabilized "PTFE" Carbon base as a single molding resulting in a totally inert pure "PTFE" inner surface. It is heatable to 270 degree C without distortion and has improved heat transfer. All "PTFE" Beaker caps can be used for Thermotech "PTFE" Beakers, accordingly to its sizes.
Height: 94 mm