Learn More
Bellco Glass, Inc. Thermowell Assembly ,15L For Vertical Over Head Drive Vessels
SDP
Catalog No. 501944791
Encompass_Preferred
Thermowell assembly is manufactured from 316l stainless steel with 32ra finish or better. It can be used with rtd temperature sensor. Includes polypropylene screw cap and silicone gasket. There are also tubes for addition media and vent. Thermowell assemblies attach to the sidearms with a screw cap assembly. 3-PORT THERMOWELL ASSY - 45mm. FOR 1964 VERTICAL OHD VESSELS. WELDED - ELECTROPOLISH FINISH. ALL MATERIAL IS: 316L SST. MADE IN THE USA. CONSIST OF:1 - SST 5/16"OD X .225"ID X 1.0" above x 15 31/32" insertionX 17 3/32" LONG 1.0" above x 15 31/32" insertion1- SST VENT TUBE; .250"OD X .152" ID)SUPPLY WITH CAP ( A523-599) & A523-600 (LINER)
Catalog No. 50-194-4791 Supplier Bellco Glass, Inc. Supplier No. 196434015R
Thermowell assembly is manufactured from 316l stainless steel with 32ra finish or better. It can be used with rtd temperature sensor. Includes polypropylene screw cap and silicone gasket. There are also tubes for addition media and vent. Thermowell assemblies attach to the sidearms with a screw cap assembly. 3-PORT THERMOWELL ASSY - 45mm. FOR 1964 VERTICAL OHD VESSELS. WELDED - ELECTROPOLISH FINISH. ALL MATERIAL IS: 316L SST. MADE IN THE USA. CONSIST OF:1 - SST 5/16"OD X .225"ID X 1.0" above x 15 31/32" insertionX 17 3/32" LONG 1.0" above x 15 31/32" insertion1- SST VENT TUBE; .250"OD X .152" ID)SUPPLY WITH CAP ( A523-599) & A523-600 (LINER)