Learn More
Bellco Glass, Inc. Thermowell Assembly ,36L For Vertical Over Head Drive Vessels
SDP
Catalog No. 501944792
Encompass_Preferred
Thermowell assembly is manufactured from 316l stainless steel with 32ra finish or better. It can be used with rtd temperature sensor. Includes polypropylene screw cap and silicone gasket. There are also tubes for addition media and vent. Thermowell assemblies attach to the sidearms with a screw cap assembly. MADE IN THE USA. 3-PORT THERMOWELL ASSY - 45mmFOR 1964 VERTICAL OHD VESSELSWELDED - ELECTROPOLISH FINISHALL MATERIAL IS: 316L SSTCONSIST OF:1 - SST 5/16"OD X .225"ID X 17 3/32" LONG 1.0" above x 17 3/32" insertion1- SST FILL TUBE; .250"OD X .152" ID (SAME DIMESIONS AS THERMOWELL TUBE)1- SST VENT TUBE: .250" OD X .152" ID X 2 5/32" LONGSUPPLY WITH CAP ( A523-599) & A523-600 (LINER)
Catalog No. 50-194-4792 Supplier Bellco Glass, Inc. Supplier No. 196434036R
Thermowell assembly is manufactured from 316l stainless steel with 32ra finish or better. It can be used with rtd temperature sensor. Includes polypropylene screw cap and silicone gasket. There are also tubes for addition media and vent. Thermowell assemblies attach to the sidearms with a screw cap assembly. MADE IN THE USA. 3-PORT THERMOWELL ASSY - 45mmFOR 1964 VERTICAL OHD VESSELSWELDED - ELECTROPOLISH FINISHALL MATERIAL IS: 316L SSTCONSIST OF:1 - SST 5/16"OD X .225"ID X 17 3/32" LONG 1.0" above x 17 3/32" insertion1- SST FILL TUBE; .250"OD X .152" ID (SAME DIMESIONS AS THERMOWELL TUBE)1- SST VENT TUBE: .250" OD X .152" ID X 2 5/32" LONGSUPPLY WITH CAP ( A523-599) & A523-600 (LINER)