Learn More
Bellco Glass, Inc. Thermowell Assembly , 6L For Angled Sidearms
SDP
Catalog No. 501945016
Encompass_Preferred
Thermowell assemblies are used to hold inside one tube the rtd temperature sensor that monitors the temperature within the vessel in conjunction with a temperature controller. There are also tubes for addition of media and vent tube. MADE IN THE USA. •Thermowell assemblies for 1965 and 1967-style angled sidearm flasks, 3-port•5/16" od thermowell tube,1/4" siphon and vent tubes•Includes 45mm pp cap and silicone gasket•Seam welded and electropolished finish - outside only•All tubes: 316l sst•Longest tube length @ curve: 10 1/2"
Catalog No. 50-194-5016 Supplier Bellco Glass, Inc. Supplier No. 196532006R
Add to Cart
Add to Cart
Thermowell assemblies are used to hold inside one tube the rtd temperature sensor that monitors the temperature within the vessel in conjunction with a temperature controller. There are also tubes for addition of media and vent tube. MADE IN THE USA. •Thermowell assemblies for 1965 and 1967-style angled sidearm flasks, 3-port•5/16" od thermowell tube,1/4" siphon and vent tubes•Includes 45mm pp cap and silicone gasket•Seam welded and electropolished finish - outside only•All tubes: 316l sst•Longest tube length @ curve: 10 1/2"