Learn More
Bellco Glass, Inc. Thermowell Assembly , 8L For Vertical Over Head Drive Vessels
SDP
Catalog No. 501944790
Encompass_Preferred
Thermowell assembly is manufactured from 316l stainless steel with 32ra finish or better. It can be used with rtd temperature sensor. Includes polypropylene screw cap and silicone gasket. There are also tubes for addition media and vent. Thermowell assemblies attach to the sidearms with a screw cap assembly. MADE IN THE USA. Specifications:Material: 316L Stainless Steel 32Ra Finish or BetterIncludes: Screw Cap & Silicone Gasket
Catalog No. 50-194-4790 Supplier Bellco Glass, Inc. Supplier No. 196434008R
Thermowell assembly is manufactured from 316l stainless steel with 32ra finish or better. It can be used with rtd temperature sensor. Includes polypropylene screw cap and silicone gasket. There are also tubes for addition media and vent. Thermowell assemblies attach to the sidearms with a screw cap assembly. MADE IN THE USA. Specifications:Material: 316L Stainless Steel 32Ra Finish or BetterIncludes: Screw Cap & Silicone Gasket