Learn More
Bellco Glass, Inc. Thermowell Fitting,5/16 in For 1/2 Hole"
SDP
Catalog No. 501946281
Encompass_Preferred
316L SST, RA FINISH OF 32 OR BETTER MICRO INCHES1/2" COMPRESSION FITT'G TO BE USED IN A 1/2" HOLE IN THE L/F HEADPLATEFOR THERMOWELLS - 5/16" TUBE. MADE IN THE USA
Catalog No. 50-194-6281 Supplier Bellco Glass, Inc. Supplier No. 780311540
316L SST, RA FINISH OF 32 OR BETTER MICRO INCHES1/2" COMPRESSION FITT'G TO BE USED IN A 1/2" HOLE IN THE L/F HEADPLATEFOR THERMOWELLS - 5/16" TUBE. MADE IN THE USA