Learn More
Chemglass Life Sciences Thermowell, M10, 300mm Immersion Length, for 7L Bio Reactors
SDP
Catalog No. 501294520
Encompass
Thermowell, M10, 300mm Immersion Length, for 7L Bio Reactors. M10 threaded thermowell having a ID of .310 that will accept 1/4 thermocouple probes. To enhance the temperature measurement the thermowell should be partially filled with a heat conducting silicone or mineral oil.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-129-4520 Supplier Chemglass Life Sciences Supplier No. CLS138305
Thermowell, M10, 300mm Immersion Length, for 7L Bio Reactors. M10 threaded thermowell having a ID of .310 that will accept 1/4 thermocouple probes. To enhance the temperature measurement the thermowell should be partially filled with a heat conducting silicone or mineral oil.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.