Learn More
Sigma Aldrich Fine Chemicals Biosciences Thermowell heating mantle1EA
Catalog No. 502900399
Encompass
Economical and dependable. These mantles have a ceramic heat cavity (resistance wire completely embedded) a grounded metal housing for safer operation a low profile that permits magnetic stirring and can be operated at up to 550 C. May be used as an air or sand bath. All models have a 36 in. 3-wire integral power cord with U.S. plug (used as an interconnector plug with POWERMITE(TM) heat controllers). 50/60AHz.
Catalog No. 50-290-0399 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2436551EA
Economical and dependable. These mantles have a ceramic heat cavity (resistance wire completely embedded) a grounded metal housing for safer operation a low profile that permits magnetic stirring and can be operated at up to 550 C. May be used as an air or sand bath. All models have a 36 in. 3-wire integral power cord with U.S. plug (used as an interconnector plug with POWERMITE(TM) heat controllers). 50/60AHz.