Learn More
Sigma Aldrich Fine Chemicals Biosciences Thermowell heating mantle1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z2436981EA
Encompass
Economical and dependable. These mantles have a ceramic heat cavity (resistance wire completely embedded), a grounded metal housing for safer operation, a low profile that permits magnetic stirring, and can be operated at up to 550 C. May be used as an air or sand bath. All models have a 36 in., 3-wire, integral power cord with U.S. plug (used as an interconnector plug with POWERMITE(TM) heat controllers). 50/60AHz.
Catalog No. 50-289-1523