Learn More
Chemglass Life Sciences Thermowell, M10, 370mm Immersion Length, for 15L Bio Reactors
SDP
Catalog No. 5000083197
Encompass
M10 threaded thermowell having a ID of .310” that will accept 1/4” thermocouple probes. To enhance the temperature measurement the thermowell should be partially filled with a heat conducting silicone or mineral oil. Stainless steel components manufactured from 316L with an electropolished surface finish of 20Ra or better.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-000-83197 Supplier Chemglass Life Sciences Supplier No. CLS138510
M10 threaded thermowell having a ID of .310” that will accept 1/4” thermocouple probes. To enhance the temperature measurement the thermowell should be partially filled with a heat conducting silicone or mineral oil. Stainless steel components manufactured from 316L with an electropolished surface finish of 20Ra or better.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.