Learn More
Bellco Glass, Inc. Thermowell Pipe Assembly , 15L For 10mm Port
SDP
Catalog No. 501946485
Encompass_Preferred
Stainless steal Thermowell holds liquid and RTD temperature sensor for recording internal bioreactor temperature. Signal is read by temperature controller and heating blanket temperature increases and decreases as required to maintain temperature setting. Thermowell Pipe Assy, Size: 15L, For 10mm Port. Diameter of Pipe: .375" OD x .305" ID - RA 20 Finish - inside & out. Length of Pipe: 16 9/16" Overall. Insertion Length: 15 3/4". Supplied with: A523-234 - #011 Silicone O-Ring, A555-23 - SSt Washer, and A555-3 - SST Nut. MADE IN THE USA
Catalog No. 50-194-6485 Supplier Bellco Glass, Inc. Supplier No. 790915089
Stainless steal Thermowell holds liquid and RTD temperature sensor for recording internal bioreactor temperature. Signal is read by temperature controller and heating blanket temperature increases and decreases as required to maintain temperature setting. Thermowell Pipe Assy, Size: 15L, For 10mm Port. Diameter of Pipe: .375" OD x .305" ID - RA 20 Finish - inside & out. Length of Pipe: 16 9/16" Overall. Insertion Length: 15 3/4". Supplied with: A523-234 - #011 Silicone O-Ring, A555-23 - SSt Washer, and A555-3 - SST Nut. MADE IN THE USA