Learn More
Bellco Glass, Inc. Thermowell Pipe Assembly , 3L For 10mm Port
SDP
Catalog No. 501946432
Encompass_Preferred
Stainless steal Thermowell holds liquid and RTD temperature sensor for recording internal bioreactor temperature. Signal is read by temperature controller and heating blanket temperature increases and decreases as required to maintain temperature setting. Fits into 10mm port on 3L metric headplate.. MADE IN THE USA
Catalog No. 50-194-6432 Supplier Bellco Glass, Inc. Supplier No. 790900021
Stainless steal Thermowell holds liquid and RTD temperature sensor for recording internal bioreactor temperature. Signal is read by temperature controller and heating blanket temperature increases and decreases as required to maintain temperature setting. Fits into 10mm port on 3L metric headplate.. MADE IN THE USA