Learn More
Bellco Glass, Inc. Thermowell Pipe Assembly , 7L For 10mm Port
SDP
Catalog No. 501946448
Encompass_Preferred
Thermowell Pipe Assy. Size: 7L. RA Finish of Assy - RA32 or better -. For 10mm Port. Diameter of Pipe: .375" OD x .305" ID - RA 20 Finish - inside & out. Length of Pipe: 12" Overall. MADE IN THE USA
Catalog No. 50-194-6448 Supplier Bellco Glass, Inc. Supplier No. 790900041
Thermowell Pipe Assy. Size: 7L. RA Finish of Assy - RA32 or better -. For 10mm Port. Diameter of Pipe: .375" OD x .305" ID - RA 20 Finish - inside & out. Length of Pipe: 12" Overall. MADE IN THE USA