Learn More
Med Vet International Tympanic Ear Thermometer Braun Thermoscan Ear Probe Handheld, 06000-300, Each
Catalog No. 502829352
Encompass_Preferred
With large two-box cradle (holds 40 covers) Pulse timer, memory recall and C/F conversion buttons 2 x 1.5 V type AA (alkaline) batteries Innovative PerfecTemp technology adjusts for variability in probe placement ExacTemp technology detects stability of the probe during measurement A pre-warmed sensor helps support accurate measurements Designed with plastics compatible with common medical-grade cleaning products Memory recall button displays last measurement taken Pre-warmed probe tip enhances patient comfort during reading The large cradle can be wall mounted for added security when used with an optional security tether (Welch Allyn number 106204, McKesson number 984426 / 16242500) sold separately Digital Thermometer works with Welch Allyn Probe Cover #06000-005 (McKesson Item 1002774 / 70052500) sold separately
Catalog No. 50-282-9352 Supplier Med Vet International Supplier No. 959367EA
With large two-box cradle (holds 40 covers) Pulse timer, memory recall and C/F conversion buttons 2 x 1.5 V type AA (alkaline) batteries Innovative PerfecTemp technology adjusts for variability in probe placement ExacTemp technology detects stability of the probe during measurement A pre-warmed sensor helps support accurate measurements Designed with plastics compatible with common medical-grade cleaning products Memory recall button displays last measurement taken Pre-warmed probe tip enhances patient comfort during reading The large cradle can be wall mounted for added security when used with an optional security tether (Welch Allyn number 106204, McKesson number 984426 / 16242500) sold separately Digital Thermometer works with Welch Allyn Probe Cover #06000-005 (McKesson Item 1002774 / 70052500) sold separately