Learn More
Med Vet International Tympanic Ear Thermometer Braun Thermoscan Ear Probe Handheld, 06000-200, Each
Catalog No. 502829350
Encompass_Preferred
With small one-box cradle (holds 20 covers) Pulse timer, memory recall and C/F conversion buttons 2 x 1.5 V type AA (alkaline) batteries Innovative PerfecTemp technology adjusts for variability in probe placement ExacTemp technology detects stability of the probe during measurement A pre-warmed sensor helps support accurate measurements Designed with plastics compatible with common medical-grade cleaning products Memory recall button displays last measurement taken Pre-warmed probe tip enhances patient comfort during reading
Catalog No. 50-282-9350 Supplier Med Vet International Supplier No. 959366EA
With small one-box cradle (holds 20 covers) Pulse timer, memory recall and C/F conversion buttons 2 x 1.5 V type AA (alkaline) batteries Innovative PerfecTemp technology adjusts for variability in probe placement ExacTemp technology detects stability of the probe during measurement A pre-warmed sensor helps support accurate measurements Designed with plastics compatible with common medical-grade cleaning products Memory recall button displays last measurement taken Pre-warmed probe tip enhances patient comfort during reading