Learn More
Research Products International Corp ThermalSeal RT2RR Films for RT qPCR, Optically Clear, Non-S, 100/pk
SDP
Catalog No. 502230139
Encompass
ThermalSeal RT Films are optically transparent, non-absorbing, non-fluorescing and designed for use in Real-Time PCR applications. Desgined specically for Raised Rim Plates these- films feature a smooth adhesive allowing extreme optical clarity and superior light transmission. End tabs and split backing allow accurate plate positioning. Available 100 per package.  ThermalSeal RT2RR is a Trademark of Excel Scientific, Inc.
Ideal for optical assay
Brilliant ultra-high optical clarity
RNase, DNase and nucleic acid free
Withstand -40° to +120 °C
Catalog No. 50-223-0139 Supplier Research Products International Corp Supplier No. 252005
ThermalSeal RT Films are optically transparent, non-absorbing, non-fluorescing and designed for use in Real-Time PCR applications. Desgined specically for Raised Rim Plates these- films feature a smooth adhesive allowing extreme optical clarity and superior light transmission. End tabs and split backing allow accurate plate positioning. Available 100 per package.  ThermalSeal RT2RR is a Trademark of Excel Scientific, Inc.
Ideal for optical assay
Brilliant ultra-high optical clarity
RNase, DNase and nucleic acid free
Withstand -40° to +120 °C