Learn More
Perkin Elmer US LLC Iron Calibration Material - Qty. 3
Catalog No. 502016381
Encompass_Preferred
PerkinElmer's calibration and reference materials ensure accurate theromgravimetric measurements for all of their TGA instruments. Magnetic calibration reference standards are available as a kit as well as individually. PerkinElmer recommends the use of a fresh reference standard every time you calibrate your instruments, to avoid the effects of hysteresis and oxidation on the Curie Point of the materials.
Catalog No. 50-201-6381 Supplier Perkin Elmer US LLC Supplier No. N5320112
PerkinElmer's calibration and reference materials ensure accurate theromgravimetric measurements for all of their TGA instruments. Magnetic calibration reference standards are available as a kit as well as individually. PerkinElmer recommends the use of a fresh reference standard every time you calibrate your instruments, to avoid the effects of hysteresis and oxidation on the Curie Point of the materials.