Learn More
Enzo Life Sciences 14-3-3 theta / tau (human), (recombinant) (His-tag) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE4870100
Encompass_Preferred
Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In 50mM NaH2PO4, pH 8.0, 300mM NaCl. Source: Produced in E. coli. Human 14-3-3τ/θ is fused at the C-terminus to a His-tag. Long Term Storage: -80°C.
Catalog No. 50-201-2048