Learn More
Aviva Systems Biology Corp THEX1 Antibody - C-terminal region (ARP41158_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41158_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against THEX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human THEX1. Alternate names: HEXO, THEX1, 3'HEXOApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57743