Learn More
Perkin Elmer US LLC Standard THGA Graphite Contact Cylinders, 5 Pairs
Catalog No. 509052005
Encompass_Preferred
The graphite contact cylinders are engineered for a precise fit so that variations in electrical contact, which might affect the analytical performance, are reduced to a minimum.
Catalog No. 50-905-2005 Supplier Perkin Elmer US LLC Supplier No. B0504036
The graphite contact cylinders are engineered for a precise fit so that variations in electrical contact, which might affect the analytical performance, are reduced to a minimum.