Learn More
Perkin Elmer US LLC Standard THGA Graphite Contact Cylinders, 1 Pair
Catalog No. 509052004
Encompass_Preferred
The graphite contact cylinders are engineered for a precise fit so that variations in electrical contact, which might affect the analytical performance, are reduced to a minimum.
Catalog No. 50-905-2004 Supplier Perkin Elmer US LLC Supplier No. B0504035
The graphite contact cylinders are engineered for a precise fit so that variations in electrical contact, which might affect the analytical performance, are reduced to a minimum.