Learn More
Cayman Chemical Thiabendazole, Purity: ≥98%, CAS: 148-79-8, 50 g
SDP
Catalog No. 502987960
Encompass_Preferred
A broad-spectrum anthelmintic active against a variety of helminths; kills greater than 95% of adult worms from ten genera; inhibits egg production and disrupts larval development; inhibits fumarate reductase in helminths, blocking energy production through the citric acid cycle; inhibits methionine aminopeptidase in E. coli (Ki = 40 nM; IC50 = 47.2 nM)
Catalog No. 50-298-7960 Supplier Cayman Chemical Supplier No. 2339150 G
A broad-spectrum anthelmintic active against a variety of helminths; kills greater than 95% of adult worms from ten genera; inhibits egg production and disrupts larval development; inhibits fumarate reductase in helminths, blocking energy production through the citric acid cycle; inhibits methionine aminopeptidase in E. coli (Ki = 40 nM; IC50 = 47.2 nM)