Learn More
Medchemexpress LLC Thiabendazole-d4 (2-(4-thiazolyl)benzimidazole-d4) | 1190007-20-5 | 98.9% | 5 MG
SDP
Catalog No. 5000405661
Encompass_Preferred
Thiabendazole-d4 is a deuterated form of Thiabendazole, an antiseptic, antifungal, and antiparasitic agent intended for research use only. This compound, appearing as a white to off-white solid, can be used as a tracer and an internal standard for quantitative analysis. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05661 Supplier Medchemexpress LLC Supplier No. HYB0263S5MG
Thiabendazole-d4 is a deuterated form of Thiabendazole, an antiseptic, antifungal, and antiparasitic agent intended for research use only. This compound, appearing as a white to off-white solid, can be used as a tracer and an internal standard for quantitative analysis. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS